Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 3 | 671 | ||
NM_004175.4 | NP_004166.1 | 3 | 685 | |||
GenBank mRNA | AB451249.1 | - | 2 | 245 | ||
AB451373.1 | - | 2 | 245 | |||
AK296173.1 | - | 3 | 328 | |||
BC000457.2 | - | 3 | 341 | 2822042 | ||
BC003150.1 | - | 3 | 341 | 2822042 | ||
BC034447.1 | - | 3 | 368 | |||
BM928709.1 | - | 3 | 633 | |||
BY993733.1 | - | 3 | 357 | |||
CN286602.1 | - | 3 | 325 | |||
CR456583.1 | - | 3 | 361 | |||
U15009.1 | - | 3 | 332 |