Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136019.2 | NP_001129491.1 | 7 | 1562 | ||
NM_004107.4 | NP_004098.1 | 6 | 1298 | |||
XM_005258657.1 | XP_005258714.1 | 6 | 1117 | |||
XM_011526627.2 | XP_011524929.1 | 5 | 979 | |||
GenBank mRNA | AK074734.1 | - | 3 | 2440 | ||
AK075510.1 | - | 7 | 1496 | |||
AK075532.1 | - | 7 | 1212 | |||
BC008734.2 | - | 6 | 1242 | 3163446 | ||
BE245284.1 | - | 3 | 332 | |||
BT006828.1 | - | 6 | 1076 | |||
BX647163.1 | - | 7 | 1182 | |||
U12255.1 | - | 6 | 1201 |