Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TROVE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042369.2 | NP_001035828.1 | 2 | 480 | ||
NM_001042370.2 | NP_001035829.2 | 2 | 480 | |||
NM_001173524.1 | NP_001166995.1 | 2 | 486 | |||
NM_001173525.1 | NP_001166996.1 | 2 | 480 | |||
NM_004600.5 | NP_004591.2 | 2 | 480 | |||
XM_006711495.3 | XP_006711558.1 | 2 | 486 | |||
XM_006711496.3 | XP_006711559.1 | 3 | 662 | |||
XM_006711497.3 | XP_006711560.1 | 3 | 662 | |||
XM_017002180.1 | XP_016857669.1 | 2 | 295 | |||
XM_017002181.1 | XP_016857670.1 | 3 | 590 | |||
XM_017002182.1 | XP_016857671.1 | 2 | 295 | |||
GenBank mRNA | AK314594.1 | - | 2 | 273 | ||
AY205314.1 | - | 2 | 294 | |||
AY205315.1 | - | 2 | 294 | |||
BC036658.2 | - | 2 | 322 | 4821776 | ||
CX761715.1 | - | 3 | 620 | |||
DA518443.1 | - | 2 | 486 | |||
J04137.1 | - | 1 | 149 | |||
M25077.1 | - | 2 | 294 |