Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270427.1 | NP_001257356.1 | 9 | 2588 | ||
NM_005047.3 | NP_005038.1 | 10 | 2717 | |||
XM_017014928.1 | XP_016870417.1 | 9 | 2517 | |||
GenBank mRNA | AK026925.1 | - | 6 | 2020 | ||
BC014478.1 | - | 10 | 2608 | 4878087 | ||
D31889.1 | - | 10 | 2604 |