Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193272.1 | NP_001180201.1 | 14 | 2543 | ||
NM_001193273.1 | NP_001180202.1 | 14 | 2598 | |||
NM_005057.3 | NP_005048.2 | 14 | 2657 | |||
XM_011509833.2 | XP_011508135.1 | 14 | 2652 | |||
GenBank mRNA | BC037284.2 | - | 14 | 2496 | 5266066 | |
BC053856.1 | - | 14 | 2628 | 5576406 | ||
X85134.1 | - | 14 | 2620 |