Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTK2 SIRNA Assays ›
Gene Symbol:
PTK2
Gene Name:
protein tyrosine kinase 2
Gene Aliases:
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location:
Chr.8: 140658382 - 141001313 on Build GRCh38
UniGene ID:
Hs.395482
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142861
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199649.1
NP_001186578.1
6
776
NM_001316342.1
NP_001303271.1
4
520
NM_005607.4
NP_005598.3
6
765
NM_153831.3
NP_722560.1
6
776
XM_005251003.2
XP_005251060.1
5
817
XM_005251012.2
XP_005251069.1
5
659
XM_005251013.2
XP_005251070.1
4
478
XM_006716608.2
XP_006716671.1
6
748
XM_006716609.2
XP_006716672.1
5
581
XM_006716610.2
XP_006716673.1
5
573
XM_011517194.2
XP_011515496.1
6
717
XM_017013651.1
XP_016869140.1
4
498
XM_017013652.1
XP_016869141.1
4
498
XM_017013653.1
XP_016869142.1
5
817
XM_017013654.1
XP_016869143.1
5
817
XM_017013655.1
XP_016869144.1
5
817
XM_017013656.1
XP_016869145.1
5
817
XM_017013657.1
XP_016869146.1
5
817
XM_017013658.1
XP_016869147.1
6
641
XM_017013659.1
XP_016869148.1
4
498
XM_017013660.1
XP_016869149.1
6
641
XM_017013661.1
XP_016869150.1
5
659
XM_017013662.1
XP_016869151.1
6
641
XM_017013663.1
XP_016869152.1
5
572
XM_017013664.1
XP_016869153.1
6
748
XM_017013665.1
XP_016869154.1
5
572
XM_017013666.1
XP_016869155.1
5
817
XM_017013667.1
XP_016869156.1
6
641
XM_017013668.1
XP_016869157.1
5
659
XM_017013669.1
XP_016869158.1
6
641
XM_017013670.1
XP_016869159.1
6
641
XM_017013671.1
XP_016869160.1
6
641
XM_017013672.1
XP_016869161.1
5
572
XM_017013673.1
XP_016869162.1
5
572
XM_017013674.1
XP_016869163.1
6
748
XM_017013675.1
XP_016869164.1
5
572
XM_017013676.1
XP_016869165.1
4
498
XM_017013677.1
XP_016869166.1
6
641
XM_017013678.1
XP_016869167.1
5
572
XM_017013679.1
XP_016869168.1
5
572
XM_017013680.1
XP_016869169.1
5
1633
XM_017013681.1
XP_016869170.1
5
581
XM_017013682.1
XP_016869171.1
4
478
XM_017013683.1
XP_016869172.1
5
581
XM_017013684.1
XP_016869173.1
5
581
XM_017013685.1
XP_016869174.1
5
525
XM_017013686.1
XP_016869175.1
4
478
XM_017013687.1
XP_016869176.1
5
581
XM_017013688.1
XP_016869177.1
5
581
XM_017013689.1
XP_016869178.1
4
446
XM_017013690.1
XP_016869179.1
4
420
GenBank mRNA
AB209071.1
-
4
426
AB209083.1
-
6
363
AK304356.1
-
6
621
AL832961.1
-
6
648
BC035404.2
-
4
497
4999680
KF437463.1
-
4
469
L13616.1
-
6
698
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
angiogenesis
vasculogenesis
neuron migration
placenta development
regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
cellular component disassembly involved in execution phase of apoptosis
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
axon guidance
positive regulation of cell proliferation
regulation of cell shape
embryo development
regulation of endothelial cell migration
regulation of epithelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
central nervous system neuron axonogenesis
negative regulation of cell-cell adhesion
establishment of cell polarity
extracellular matrix organization
positive regulation of cell migration
regulation of cell adhesion mediated by integrin
netrin-activated signaling pathway
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
establishment of nucleus localization
regulation of cell proliferation
negative regulation of apoptotic process
regulation of GTPase activity
endothelial cell migration
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
innate immune response
regulation of osteoblast differentiation
positive regulation of protein kinase activity
negative regulation of organ growth
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
cell motility
negative regulation of axonogenesis
regulation of cytoskeleton organization
regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of synapse assembly
growth hormone receptor signaling pathway
regulation of substrate adhesion-dependent cell spreading
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of anoikis
actin binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
protein binding
ATP binding
JUN kinase binding
protein kinase binding
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-nsq4p:80/10.239.163.53:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0