Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025070.1 | NP_001020241.1 | 3 | 464 | ||
NM_001025071.1 | NP_001020242.1 | 3 | 470 | |||
NM_005617.3 | NP_005608.1 | 3 | 253 | |||
XM_017009684.1 | XP_016865173.1 | 3 | 307 | |||
GenBank mRNA | AA467888.1 | - | 1 | 20 | ||
AK312179.1 | - | 3 | 232 | |||
BC001126.1 | - | 1 | 216 | 2967098 | ||
BC003401.1 | - | 2 | 210 | 3448752 | ||
BC006784.1 | - | 1 | 221 | 2900907 | ||
BC020515.1 | - | 3 | 403 | 3856643 | ||
BC091474.1 | - | 3 | 234 | 30334486 | ||
BM454896.1 | - | 3 | 433 | |||
BM557014.1 | - | 3 | 405 |
Gene Symbol: RPS14P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077246.1 | 1 | 504 | |||
GenBank mRNA | BC072682.1 | - | 1 | 504 | ||
BQ930453.1 | - | 1 | 506 | |||
BU176905.1 | - | 1 | 472 |