Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006743.4 | NP_006734.1 | 7 | 766 | ||
GenBank mRNA | AK000859.1 | - | 7 | 635 | ||
AK026664.1 | - | 3 | 1093 | |||
AK309137.1 | - | 7 | 1024 | |||
AY203954.1 | - | 6 | 1228 | |||
BC006825.1 | - | 7 | 595 | 3449185 | ||
BQ429805.1 | - | 6 | 530 | |||
BX647914.1 | - | 1 | 2986 |