Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 8, 9 | 1328 | ||
NM_173701.1 | NP_776049.1 | 8, 9 | 1104 | |||
NM_213645.1 | NP_998810.1 | 7, 8 | 932 | |||
NM_213646.1 | NP_998811.1 | 7, 8 | 1156 | |||
XM_005268044.3 | XP_005268101.1 | 8, 9 | 1056 | |||
XM_006720249.3 | XP_006720312.1 | 9, 10 | 1290 | |||
XM_011537133.2 | XP_011535435.1 | 10, 11 | 1302 | |||
XM_011537135.2 | XP_011535437.1 | 9, 10 | 1186 | |||
XM_011537136.2 | XP_011535438.1 | 8, 9 | 1022 | |||
XM_017021627.1 | XP_016877116.1 | 9, 10 | 1194 | |||
XM_017021628.1 | XP_016877117.1 | 9, 10 | 1325 | |||
XM_017021629.1 | XP_016877118.1 | 10, 11 | 1300 | |||
GenBank mRNA | AK025487.1 | - | 6, 7 | 732 | ||
AK056100.1 | - | 8, 9 | 1566 | |||
AK130698.1 | - | 4, 5 | 435 | |||
AK291141.1 | - | 7, 8 | 908 | |||
BC017489.1 | - | 8, 9 | 1204 | 3542671 | ||
BC095453.1 | - | 9, 10 | 1215 | 30418785 | ||
BX248006.1 | - | 8, 9 | 1048 | |||
M61715.1 | - | 8, 9 | 1134 | |||
M77804.1 | - | 7, 8 | 989 | |||
X59892.1 | - | 8, 9 | 1058 | |||
X62570.1 | - | 7, 8 | 1009 |