Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006902.4 | NP_008833.1 | 5 | 907 | ||
NM_022716.3 | NP_073207.1 | 4 | 835 | |||
XM_006711388.3 | XP_006711451.1 | 5 | 898 | |||
GenBank mRNA | BI039001.1 | - | 2 | 321 |