Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143165
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
6
621
NM_001318068.1
NP_001304997.1
6
716
NM_001318069.1
NP_001304998.1
7
1186
NM_002753.4
NP_002744.1
7
722
NM_138980.3
NP_620446.1
7
1213
NM_138982.3
NP_620448.1
7
1186
XM_005263129.2
XP_005263186.1
7
1283
XM_005263130.2
XP_005263187.1
7
679
XM_005263131.2
XP_005263188.1
7
1080
XM_005263135.3
XP_005263192.1
7
1080
XM_006714268.2
XP_006714331.1
7
953
XM_006714269.2
XP_006714332.1
7
1080
XM_011532117.2
XP_011530419.1
7
786
XM_011532118.2
XP_011530420.1
7
879
XM_011532120.2
XP_011530422.1
7
849
XM_011532121.2
XP_011530423.1
7
715
XM_017008419.1
XP_016863908.1
8
976
XM_017008420.1
XP_016863909.1
7
1410
XM_017008421.1
XP_016863910.1
7
1006
XM_017008422.1
XP_016863911.1
5
506
XM_017008423.1
XP_016863912.1
7
698
XM_017008424.1
XP_016863913.1
7
1033
XM_017008425.1
XP_016863914.1
7
706
XM_017008426.1
XP_016863915.1
7
1034
XM_017008427.1
XP_016863916.1
7
1107
XM_017008428.1
XP_016863917.1
7
849
XM_017008429.1
XP_016863918.1
7
1410
XM_017008430.1
XP_016863919.1
7
788
XM_017008431.1
XP_016863920.1
7
1006
XM_017008432.1
XP_016863921.1
7
679
XM_017008433.1
XP_016863922.1
7
1107
XM_017008434.1
XP_016863923.1
7
1063
XM_017008435.1
XP_016863924.1
7
953
XM_017008436.1
XP_016863925.1
7
1437
XM_017008437.1
XP_016863926.1
7
715
XM_017008438.1
XP_016863927.1
7
706
XM_017008439.1
XP_016863928.1
7
1034
XM_017008440.1
XP_016863929.1
7
1107
XM_017008441.1
XP_016863930.1
7
1107
XM_017008442.1
XP_016863931.1
9
1428
XM_017008443.1
XP_016863932.1
9
1079
XM_017008444.1
XP_016863933.1
9
1401
XM_017008445.1
XP_016863934.1
6
910
XM_017008446.1
XP_016863935.1
4
1602
XM_017008447.1
XP_016863936.1
6
1048
XM_017008448.1
XP_016863937.1
6
1021
XM_017008449.1
XP_016863938.1
6
1048
XM_017008450.1
XP_016863939.1
6
1021
XM_017008451.1
XP_016863940.1
6
847
XM_017008452.1
XP_016863941.1
6
1048
XM_017008453.1
XP_016863942.1
5
506
XM_017008454.1
XP_016863943.1
7
786
GenBank mRNA
AK057723.1
-
2
508
AK124791.1
-
7
1438
AK225974.1
-
7
767
AK294125.1
-
3
237
AK307129.1
-
7
840
BC022492.1
-
7
1019
BC035057.1
-
7
1031
5261221
BC051731.1
-
7
832
4838495
BC065516.1
-
2
183
5721954
U07620.1
-
7
723
U34819.1
-
6
567
U34820.1
-
6
591
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0