Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524134.2 | XP_011522436.2 | 5 | 492 |
Gene Symbol: MAP2K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316332.1 | NP_001303261.1 | 13 | 1511 | ||
NM_002756.4 | NP_002747.2 | 12 | 1128 | |||
NM_145109.2 | NP_659731.1 | 12 | 1386 | |||
XM_005256723.2 | XP_005256780.1 | 13 | 1426 | |||
XM_011523958.2 | XP_011522260.1 | 11 | 1045 | |||
XM_011523959.1 | XP_011522261.1 | 14 | 1393 | |||
XM_017024857.1 | XP_016880346.1 | 12 | 1255 | |||
XM_017024858.1 | XP_016880347.1 | 13 | 1243 | |||
XM_017024859.1 | XP_016880348.1 | 14 | 1583 | |||
GenBank mRNA | AK093838.1 | - | 13 | 1430 | ||
BC032478.1 | - | 12 | 1321 | 5215093 | ||
D87116.1 | - | 12 | 1178 | |||
L36719.1 | - | 14 | 1371 | |||
U66839.1 | - | 12 | 1247 |