Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000434.3 | NP_000425.1 | 6 | 1710 | ||
GenBank mRNA | AB209011.1 | - | 3 | 2862 | ||
AF040958.1 | - | 6 | 1682 | |||
BC000722.2 | - | 6 | 1651 | 3506824 | ||
BC011900.2 | - | 6 | 1582 | 4101695 | ||
U84246.1 | - | 6 | 1608 | |||
X78687.1 | - | 6 | 1620 |