Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000913.5 | NP_000904.1 | 4 | 1624 | ||
NM_001200019.1 | NP_001186948.1 | 6 | 1732 | |||
NM_001318853.1 | NP_001305782.1 | 5 | 1775 | |||
NM_001318854.1 | NP_001305783.1 | 3 | 1338 | |||
NM_001318855.1 | NP_001305784.1 | 3 | 1381 | |||
NM_182647.3 | NP_872588.1 | 5 | 1775 | |||
XM_011528828.2 | XP_011527130.1 | 5 | 2277 | |||
XM_011528830.2 | XP_011527132.1 | 4 | 2012 | |||
XM_017027852.1 | XP_016883341.1 | 4 | 1794 | |||
XM_017027853.1 | XP_016883342.1 | 5 | 1714 | |||
XM_017027854.1 | XP_016883343.1 | 4 | 1649 | |||
XM_017027855.1 | XP_016883344.1 | 5 | 1741 | |||
GenBank mRNA | BC038433.1 | - | 6 | 1732 | 5164017 | |
U30185.1 | - | 4 | 1437 | |||
X77130.1 | - | 4 | 1497 |
Set Membership: |
![]() |