- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002363.4 | NP_002354.2 | 4 | 1434 | ||
NM_177404.2 | NP_796379.1 | 2 | 1277 | |||
NM_177415.2 | NP_803134.1 | 3 | 1351 | |||
GenBank mRNA | BC013772.1 | - | 2 | 1225 | 3917511 | |
X82539.1 | - | 4 | 1437 |