Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002365.4 | NP_002356.2 | 5 | 2249 | ||
XM_011545513.2 | XP_011543815.1 | 4 | 1856 | |||
XM_011545514.2 | XP_011543816.1 | 3 | 1823 | |||
GenBank mRNA | AK057441.1 | - | 5 | 2249 |