Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK3 SIRNA Assays ›
Gene Symbol:
MARK3
Gene Name:
microtubule affinity regulating kinase 3
Gene Aliases:
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location:
Chr.14: 103385364 - 103503831 on Build GRCh38
UniGene ID:
Hs.35828
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143504
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128918.2
NP_001122390.2
11
1749
NM_001128919.2
NP_001122391.2
11
1749
NM_001128920.2
NP_001122392.2
11
1749
NM_001128921.2
NP_001122393.2
10
1512
NM_002376.6
NP_002367.5
11
1749
XM_005267641.4
XP_005267698.2
11
1128
XM_005267642.4
XP_005267699.2
11
1749
XM_005267643.4
XP_005267700.2
11
1749
XM_006720146.3
XP_006720209.1
11
1123
XM_011536769.2
XP_011535071.1
10
862
XM_017021291.1
XP_016876780.1
11
1749
XM_017021292.1
XP_016876781.1
11
1749
XM_017021293.1
XP_016876782.1
12
1224
XM_017021294.1
XP_016876783.1
10
1498
XM_017021295.1
XP_016876784.1
10
1513
XM_017021296.1
XP_016876785.1
10
1530
XM_017021297.1
XP_016876786.1
10
1506
XM_017021298.1
XP_016876787.1
10
1506
XM_017021299.1
XP_016876788.1
10
1557
XM_017021300.1
XP_016876789.1
9
983
XM_017021301.1
XP_016876790.1
9
1138
XM_017021302.1
XP_016876791.1
9
983
XM_017021303.1
XP_016876792.1
9
983
XM_017021304.1
XP_016876793.1
9
983
XM_017021305.1
XP_016876794.1
9
983
XM_017021306.1
XP_016876795.1
10
1678
XM_017021307.1
XP_016876796.1
9
1629
XM_017021308.1
XP_016876797.1
10
1700
XM_017021309.1
XP_016876798.1
9
961
XM_017021310.1
XP_016876799.1
9
961
XM_017021311.1
XP_016876800.1
8
912
GenBank mRNA
AF170723.1
-
2
186
AF387637.1
-
11
1101
AF465413.1
-
11
1593
AK296643.1
-
4
386
AK307448.1
-
11
1128
BC024773.1
-
11
1572
5138958
BX161395.1
-
10
1442
BX248250.1
-
2
119
M80359.1
-
11
1254
U64205.1
-
11
1458
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
establishment of cell polarity
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0