Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002377.2 | NP_002368.1 | 1 | 310 | ||
GenBank mRNA | BC069142.1 | - | 1 | 311 | 7217001 | |
BC069581.1 | - | 1 | 310 | 7262257 | ||
BC110454.2 | - | 1 | 310 | 40016114 | ||
CR542261.1 | - | 1 | 296 | |||
M13150.1 | - | 1 | 563 |