Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002381.4 | NP_002372.1 | 3 | 951 | ||
GenBank mRNA | AJ001047.1 | - | 2 | 339 | ||
AJ224741.1 | - | 3 | 905 | |||
AK290856.1 | - | 3 | 951 | |||
BC139907.1 | - | 3 | 909 | 40147495 | ||
Y13341.1 | - | 2 | 361 |