Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205254.1 | NP_001192183.1 | 3 | 512 | ||
NM_002538.3 | NP_002529.1 | 3 | 742 | |||
XM_017008913.1 | XP_016864402.1 | 3 | 512 | |||
XM_017008914.1 | XP_016864403.1 | 3 | 487 | |||
GenBank mRNA | AB451306.1 | - | 2 | 306 | ||
AB451437.1 | - | 2 | 306 | |||
AK290697.1 | - | 3 | 474 | |||
AK313501.1 | - | 3 | 485 | |||
BC029886.1 | - | 3 | 484 | 5179203 | ||
DB226845.1 | - | 3 | 512 | |||
GQ402517.1 | - | 2 | 350 | |||
U49184.1 | - | 3 | 473 | |||
U53823.1 | - | 3 | 743 |