Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202431.1 | NP_001189360.1 | 6 | 835 | ||
NM_002574.3 | NP_002565.1 | 6 | 1052 | |||
NM_181696.2 | NP_859047.1 | 6 | 1026 | |||
NM_181697.2 | NP_859048.1 | 6 | 832 | |||
GenBank mRNA | AK131049.1 | - | 6 | 780 | ||
BC007063.1 | - | 6 | 778 | 3961375 | ||
BC021683.1 | - | 6 | 784 | 3681912 | ||
L19184.1 | - | 6 | 778 | |||
X67951.1 | - | 6 | 771 |