Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004314.2 | NP_004305.2 | 3 | 496 | ||
XM_011520114.2 | XP_011518416.1 | 4 | 669 | |||
XM_017017763.1 | XP_016873252.1 | 4 | 690 | |||
GenBank mRNA | AJ291430.1 | - | 1 | 32 | ||
BC069102.1 | - | 3 | 496 | 7216917 | ||
BC111729.1 | - | 3 | 472 | 40031715 | ||
S74683.1 | - | 3 | 496 |