Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127891.2 | NP_001121363.1 | 13 | 2279 | ||
NM_001302508.1 | NP_001289437.1 | 13 | 2093 | |||
NM_001302509.1 | NP_001289438.1 | 13 | 2022 | |||
NM_001302510.1 | NP_001289439.1 | 13 | 2015 | |||
NM_004530.5 | NP_004521.1 | 13 | 2421 | |||
GenBank mRNA | AK310314.1 | - | 13 | 2279 | ||
BC002576.2 | - | 13 | 2376 | 3161383 | ||
J03210.1 | - | 13 | 2063 |