Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288772.1 | NP_001275701.1 | 3 | 843 | ||
NM_001288774.1 | NP_001275703.1 | 3 | 836 | |||
NM_004570.5 | NP_004561.3 | 3 | 843 | |||
XM_011520696.2 | XP_011518998.1 | 3 | 971 | |||
XM_011520697.2 | XP_011518999.1 | 3 | 970 | |||
XM_011520700.2 | XP_011519002.1 | 3 | 962 | |||
XM_011520701.2 | XP_011519003.1 | 3 | 962 | |||
XM_017019470.1 | XP_016874959.1 | 3 | 971 | |||
XM_017019471.1 | XP_016874960.1 | 3 | 908 | |||
XM_017019472.1 | XP_016874961.1 | 4 | 1136 | |||
XM_017019473.1 | XP_016874962.1 | 3 | 1977 | |||
XM_017019474.1 | XP_016874963.1 | 3 | 971 | |||
XM_017019475.1 | XP_016874964.1 | 3 | 970 | |||
XM_017019476.1 | XP_016874965.1 | 3 | 972 | |||
XM_017019477.1 | XP_016874966.1 | 2 | 158 | |||
XM_017019478.1 | XP_016874967.1 | 3 | 962 | |||
GenBank mRNA | AJ000008.1 | - | 3 | 868 | ||
BC130277.1 | - | 2 | 762 | 40146308 | ||
BC144133.1 | - | 2 | 758 | |||
BC144134.1 | - | 2 | 762 | 9052654 |