Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDLRAD4 SIRNA Assays ›
Gene Symbol:
LDLRAD4
Gene Name:
low density lipoprotein receptor class A domain containing 4
Gene Aliases:
C18orf1
Chromosome Location:
Chr.18: 13217691 - 13652754 on Build GRCh38
UniGene ID:
Hs.731853
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14398
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDLRAD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003674.3
NP_001003674.1
4
2027
NM_001003675.3
NP_001003675.1
3
1973
NM_001276249.1
NP_001263178.1
4
1017
NM_001276251.1
NP_001263180.1
3
1561
NM_181481.4
NP_852146.1
6
1363
NM_181482.4
NP_852147.1
6
1568
XM_005258140.1
XP_005258197.1
7
1592
XM_006722353.1
XP_006722416.1
8
1649
XM_006722354.1
XP_006722417.1
6
1453
XM_011525737.1
XP_011524039.1
6
1451
XM_011525738.1
XP_011524040.1
7
1534
XM_017025961.1
XP_016881450.1
7
1572
XM_017025962.1
XP_016881451.1
6
1518
XM_017025963.1
XP_016881452.1
6
1305
XM_017025964.1
XP_016881453.1
6
1306
XM_017025965.1
XP_016881454.1
10
1959
XM_017025966.1
XP_016881455.1
9
1834
XM_017025967.1
XP_016881456.1
9
1831
XM_017025968.1
XP_016881457.1
8
1738
XM_017025969.1
XP_016881458.1
5
2419
XM_017025970.1
XP_016881459.1
5
880
XM_017025971.1
XP_016881460.1
4
1509
XM_017025972.1
XP_016881461.1
3
1482
GenBank mRNA
AF009424.1
-
6
1366
AF009425.1
-
5
1312
AF009426.1
-
3
965
AF009427.1
-
2
911
AK055028.1
-
4
2026
AK291574.1
-
4
889
BC029958.1
-
3
1561
BC030199.1
-
6
1502
5199354
BC066971.1
-
4
936
4794097
BM715555.1
-
1
297
BX114947.1
-
3
717
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of epithelial to mesenchymal transition
negative regulation of SMAD protein complex assembly
negative regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of pathway-restricted SMAD protein phosphorylation
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0