Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005563.3 | NP_005554.1 | 5 | 626 | ||
NM_203399.1 | NP_981944.1 | 5 | 602 | |||
NM_203401.1 | NP_981946.1 | 5 | 814 | |||
GenBank mRNA | AB209282.1 | - | 4 | 2488 | ||
AK054594.1 | - | 6 | 522 | |||
AV714035.1 | - | 6 | 571 | |||
BC014353.1 | - | 5 | 595 | 3683516 | ||
BC082228.1 | - | 4 | 524 | 2822803 | ||
BF686314.1 | - | 654 | ||||
BG030679.1 | - | 5 | 597 | |||
BG493669.1 | - | 5 | 629 | |||
BM563894.1 | - | 5 | 647 | |||
BX647885.1 | - | 4 | 2502 | |||
J04991.1 | - | 5 | 553 | |||
X53305.1 | - | 5 | 531 | |||
Z11566.1 | - | 5 | 541 |