Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134673.3 | NP_001128145.1 | 6 | 1362 | ||
NM_001145511.1 | NP_001138983.1 | 6 | 1206 | |||
NM_001145512.1 | NP_001138984.1 | 7 | 1095 | |||
NM_005595.4 | NP_005586.1 | 6 | 1362 | |||
XM_011541512.2 | XP_011539814.1 | 6 | 1423 | |||
XM_011541514.2 | XP_011539816.1 | 6 | 1356 | |||
XM_011541515.2 | XP_011539817.1 | 6 | 1423 | |||
XM_017001362.1 | XP_016856851.1 | 6 | 1284 | |||
XM_017001363.1 | XP_016856852.1 | 5 | 799 | |||
GenBank mRNA | AB037860.1 | - | 6 | 1362 | ||
AK299289.1 | - | 6 | 1206 | |||
AK299356.1 | - | 6 | 1103 | |||
AK299579.1 | - | 7 | 1095 | |||
BC022264.1 | - | 6 | 1049 | 4270934 |