Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242385.1 | NP_001229314.1 | 11 | 1735 | ||
NM_001242957.2 | NP_001229886.1 | 10 | 1661 | |||
NM_005906.5 | NP_005897.1 | 10 | 1661 | |||
NR_134935.1 | 12 | 1819 | ||||
NR_134936.1 | 11 | 1803 | ||||
XM_011514619.2 | XP_011512921.1 | 11 | 1800 | |||
XM_011514620.2 | XP_011512922.1 | 11 | 2587 | |||
XM_011514621.2 | XP_011512923.1 | 10 | 1783 | |||
XM_011514622.2 | XP_011512924.1 | 11 | 2112 | |||
XM_011514624.2 | XP_011512926.1 | 5 | 756 | |||
XM_017010863.1 | XP_016866352.1 | 11 | 2113 | |||
XM_017010864.1 | XP_016866353.1 | 11 | 2111 | |||
XM_017010865.1 | XP_016866354.1 | 10 | 1783 | |||
GenBank mRNA | AB593146.1 | - | 11 | 1689 | ||
AF505623.1 | - | 10 | 1661 | |||
AK314408.1 | - | 11 | 1800 | |||
BC039825.1 | - | 11 | 1773 | 5167621 | ||
JN226411.1 | - | 10 | 1432 |