Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040000.2 | NP_001035089.1 | 3 | 478 | ||
NM_001207008.1 | NP_001193937.1 | 3 | 478 | |||
NM_001291964.1 | NP_001278893.1 | 3 | 532 | |||
XM_005266996.3 | XP_005267053.1 | 3 | 597 | |||
XM_005266997.2 | XP_005267054.1 | 3 | 595 | |||
XM_006715491.2 | XP_006715554.1 | 3 | 595 | |||
XM_006715494.3 | XP_006715557.1 | 3 | 580 | |||
XM_011535842.2 | XP_011534144.1 | 3 | 597 | |||
XM_017010878.1 | XP_016866367.1 | 3 | 595 | |||
XM_017010879.1 | XP_016866368.1 | 3 | 597 | |||
XM_017010880.1 | XP_016866369.1 | 3 | 579 | |||
XM_017010881.1 | XP_016866370.1 | 3 | 580 | |||
XM_017010882.1 | XP_016866371.1 | 3 | 595 | |||
XM_017010883.1 | XP_016866372.1 | 3 | 595 | |||
XM_017010884.1 | XP_016866373.1 | 3 | 597 | |||
XM_017010885.1 | XP_016866374.1 | 3 | 580 | |||
XM_017010886.1 | XP_016866375.1 | 3 | 595 | |||
GenBank mRNA | AB209420.1 | - | 3 | 301 | ||
AB621809.1 | - | 3 | 532 | |||
AI271860.1 | - | 3 | 481 | |||
BE645141.1 | - | 3 | 478 | |||
U02478.1 | - | 3 | 336 |