Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005941.4 | NP_005932.2 | 4 | 828 | ||
GenBank mRNA | AB009303.1 | - | 4 | 658 | ||
AK314274.1 | - | 4 | 828 | |||
AL832595.1 | - | 5 | 867 | |||
BC069500.1 | - | 4 | 615 | 7262415 | ||
BC075004.2 | - | 4 | 572 | 30915400 | ||
BC075005.2 | - | 4 | 572 | 30915613 | ||
D83646.1 | - | 4 | 546 | |||
D83647.1 | - | 4 | 546 | |||
D85511.1 | - | 4 | 581 |