Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286561.1 | NP_001273490.1 | 8 | 740 | ||
NM_006185.3 | NP_006176.2 | 7 | 642 | |||
NR_104476.1 | 7 | 561 | ||||
XM_006718564.1 | XP_006718627.1 | 6 | 457 | |||
XM_011545054.1 | XP_011543356.1 | 6 | 457 | |||
XM_011545055.1 | XP_011543357.1 | 7 | 635 | |||
XM_011545056.2 | XP_011543358.1 | 8 | 629 | |||
XM_011545057.2 | XP_011543359.1 | 8 | 619 | |||
XM_011545058.1 | XP_011543360.1 | 6 | 656 | |||
XM_011545059.2 | XP_011543361.1 | 8 | 621 | |||
XM_011545060.2 | XP_011543362.1 | 8 | 615 | |||
XM_011545061.2 | XP_011543363.1 | 6 | 475 | |||
XM_011545062.2 | XP_011543364.1 | 6 | 488 | |||
XM_011545063.2 | XP_011543365.1 | 6 | 455 | |||
XM_011545064.1 | XP_011543366.1 | 6 | 442 | |||
XM_011545065.1 | XP_011543367.1 | 6 | 457 | |||
XM_011545066.1 | XP_011543368.1 | 6 | 457 | |||
XM_017017830.1 | XP_016873319.1 | 7 | 497 | |||
XM_017017831.1 | XP_016873320.1 | 6 | 493 | |||
GenBank mRNA | AB210007.1 | - | 7 | 443 | ||
AB621824.1 | - | 8 | 624 | |||
AK290509.1 | - | 6 | 457 | |||
AK307577.1 | - | 7 | 460 | |||
AK309041.1 | - | 6 | 464 | |||
BC004165.2 | - | 7 | 476 | |||
BC027493.1 | - | 7 | 443 | |||
BC043499.1 | - | 7 | 443 | |||
BC103765.1 | - | 7 | 561 | |||
Z11583.1 | - | 7 | 460 | |||
Z11584.1 | - | 8 | 562 |