Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144220
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
5
797
NM_001315508.1
NP_001302437.1
5
797
NM_006197.3
NP_006188.3
5
797
XM_006716336.3
XP_006716399.1
5
870
XM_006716340.3
XP_006716403.1
5
870
XM_006716349.3
XP_006716412.1
5
870
XM_011544528.2
XP_011542830.1
5
870
XM_011544529.2
XP_011542831.1
7
1176
XM_011544530.2
XP_011542832.1
6
992
XM_011544532.2
XP_011542834.1
5
870
XM_011544533.2
XP_011542835.1
5
870
XM_011544534.2
XP_011542836.1
5
870
XM_011544535.2
XP_011542837.1
5
870
XM_011544540.2
XP_011542842.1
6
990
XM_017013472.1
XP_016868961.1
7
1175
XM_017013473.1
XP_016868962.1
7
1174
XM_017013474.1
XP_016868963.1
5
870
XM_017013475.1
XP_016868964.1
7
1006
XM_017013476.1
XP_016868965.1
4
590
XM_017013477.1
XP_016868966.1
7
1174
XM_017013478.1
XP_016868967.1
7
1173
XM_017013479.1
XP_016868968.1
7
1174
XM_017013480.1
XP_016868969.1
5
870
XM_017013481.1
XP_016868970.1
7
1173
XM_017013482.1
XP_016868971.1
5
891
XM_017013483.1
XP_016868972.1
7
1172
XM_017013484.1
XP_016868973.1
4
598
XM_017013485.1
XP_016868974.1
7
1173
XM_017013486.1
XP_016868975.1
5
870
XM_017013487.1
XP_016868976.1
5
870
XM_017013488.1
XP_016868977.1
5
870
XM_017013489.1
XP_016868978.1
7
1173
XM_017013490.1
XP_016868979.1
7
1173
XM_017013491.1
XP_016868980.1
5
870
XM_017013492.1
XP_016868981.1
5
870
XM_017013493.1
XP_016868982.1
7
1173
XM_017013494.1
XP_016868983.1
5
870
XM_017013495.1
XP_016868984.1
7
1010
XM_017013496.1
XP_016868985.1
4
593
XM_017013497.1
XP_016868986.1
5
870
XM_017013498.1
XP_016868987.1
5
870
XM_017013499.1
XP_016868988.1
7
1010
XM_017013500.1
XP_016868989.1
5
870
XM_017013501.1
XP_016868990.1
7
1172
XM_017013502.1
XP_016868991.1
4
593
XM_017013503.1
XP_016868992.1
5
870
XM_017013504.1
XP_016868993.1
5
891
XM_017013505.1
XP_016868994.1
7
1010
XM_017013506.1
XP_016868995.1
5
870
XM_017013507.1
XP_016868996.1
7
1174
XM_017013508.1
XP_016868997.1
6
814
GenBank mRNA
AK307583.1
-
5
664
AX785390.1
-
3
375
BC000453.2
-
5
655
BC027477.1
-
4
576
BC065022.1
-
5
648
BC133052.1
-
4
450
8381608
BC140946.1
-
5
626
9056758
L27841.1
-
5
784
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00196390_m1
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0