Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170460.1 | NP_001163931.1 | 16 | 2987 | ||
NM_006201.4 | NP_006192.1 | 16 | 3122 | |||
NM_033018.3 | NP_148978.2 | 16 | 3229 | |||
XM_011543920.2 | XP_011542222.1 | 16 | 2710 | |||
XM_011543921.1 | XP_011542223.1 | 17 | 3105 | |||
XM_011543922.1 | XP_011542224.1 | 17 | 3093 | |||
XM_011543923.1 | XP_011542225.1 | 16 | 3023 | |||
XM_011543924.1 | XP_011542226.1 | 16 | 2833 | |||
XM_011543925.2 | XP_011542227.1 | 17 | 2915 | |||
XM_011543926.1 | XP_011542228.1 | 16 | 3876 | |||
XM_011543927.2 | XP_011542229.1 | 17 | 2953 | |||
XM_011543928.1 | XP_011542230.1 | 17 | 3308 | |||
XM_017029569.1 | XP_016885058.1 | 17 | 3394 | |||
XM_017029570.1 | XP_016885059.1 | 17 | 3204 | |||
XM_017029571.1 | XP_016885060.1 | 16 | 4165 | |||
XM_017029572.1 | XP_016885061.1 | 17 | 3242 | |||
XM_017029573.1 | XP_016885062.1 | 17 | 3597 | |||
GenBank mRNA | AB209465.1 | - | 13 | 3359 | ||
BC006190.2 | - | 16 | 2940 | 3830397 | ||
BC015607.2 | - | 16 | 3091 | 4762081 |