Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318942.1 | NP_001305871.1 | 4 | 671 | ||
NM_001318943.1 | NP_001305872.1 | 4 | 1052 | |||
NM_014623.3 | NP_055438.1 | 4 | 714 | |||
XM_005249121.3 | XP_005249178.1 | 4 | 779 | |||
GenBank mRNA | BC001754.1 | - | 4 | 672 | 3355279 | |
BM450289.1 | - | 615 | ||||
BQ232807.1 | - | 4 | 714 | |||
L10400.1 | - | 4 | 647 | |||
M27937.1 | - | 4 | 647 |