Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135824.1 | 2 | 314 | |||
GenBank mRNA | AK307907.1 | - | 2 | 278 | ||
AK311553.1 | - | 1 | 529 | |||
DA379333.1 | - | 2 | 314 |
Gene Symbol: MST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020998.3 | NP_066278.3 | 8 | 1064 | ||
XM_006713166.1 | XP_006713229.1 | 8 | 1825 | |||
XM_011533732.1 | XP_011532034.1 | 8 | 1825 | |||
XM_011533737.2 | XP_011532039.1 | 8 | 1829 | |||
XM_011533738.2 | XP_011532040.1 | 8 | 1829 | |||
XM_017006460.1 | XP_016861949.1 | 8 | 1825 | |||
XM_017006461.1 | XP_016861950.1 | 8 | 1825 | |||
XM_017006462.1 | XP_016861951.1 | 8 | 1825 | |||
XM_017006463.1 | XP_016861952.1 | 8 | 1825 | |||
XM_017006464.1 | XP_016861953.1 | 8 | 1825 | |||
XM_017006465.1 | XP_016861954.1 | 8 | 1825 | |||
XM_017006466.1 | XP_016861955.1 | 8 | 1825 | |||
XM_017006467.1 | XP_016861956.1 | 8 | 1825 | |||
XM_017006468.1 | XP_016861957.1 | 8 | 1829 | |||
GenBank mRNA | AK222893.1 | - | 8 | 955 | ||
AK294324.1 | - | 8 | 1064 | |||
AK296192.1 | - | 7 | 859 | |||
AK298396.1 | - | 8 | 970 | |||
AK298452.1 | - | 7 | 824 | |||
AK309567.1 | - | 8 | 1035 | |||
BC044862.1 | - | 6 | 2590 | |||
BC048330.1 | - | 8 | 963 | 5190966 | ||
L11924.1 | - | 8 | 960 | |||
M74178.1 | - | 8 | 950 |
Gene Symbol: MST1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271733.1 | NP_001258662.1 | 7 | 859 | ||
GenBank mRNA | AY192149.1 | - | 7 | 859 |
Gene Symbol: MST1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027504.1 | 7 | 883 | |||
GenBank mRNA | AK125231.1 | - | 2 | 314 | ||
AK293358.1 | - | 7 | 844 | |||
AK311393.1 | - | 5 | 1204 |