Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201362.1 | NP_001188291.1 | 5 | 370 | ||
NM_001201363.1 | NP_001188292.1 | 5 | 370 | |||
NM_001318884.1 | NP_001305813.1 | 5 | 370 | |||
NM_001318885.1 | NP_001305814.1 | 4 | 319 | |||
NM_001318886.1 | NP_001305815.1 | 5 | 370 | |||
NM_001318887.1 | NP_001305816.1 | 5 | 370 | |||
NM_004242.3 | NP_004233.1 | 5 | 370 | |||
NM_138730.2 | NP_620058.1 | 5 | 370 | |||
GenBank mRNA | AF274949.1 | - | 5 | 289 | ||
AF401520.1 | - | 5 | 261 | |||
AK124181.1 | - | 1 | 710 | |||
AK315391.1 | - | 5 | 314 | |||
AY043282.1 | - | 5 | 337 | |||
BC009529.2 | - | 5 | 298 | 3890883 | ||
BF790582.1 | - | 5 | 266 | |||
BI551210.1 | - | 5 | 377 | |||
BM927225.1 | - | 5 | 246 | |||
BX648085.1 | - | 1 | 3612 | |||
CF553005.1 | - | 5 | 234 | |||
CX761444.1 | - | 5 | 276 | |||
L40357.1 | - | 5 | 255 | |||
W19124.1 | - | 4 | 213 |