Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144387
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
14
1860
NM_002374.3
NP_002365.3
14
5461
NM_031845.2
NP_114033.2
11
1393
NM_031847.2
NP_114035.2
12
1486
XM_005246555.1
XP_005246612.1
17
6090
XM_005246565.1
XP_005246622.1
13
5379
XM_005246566.1
XP_005246623.1
13
5367
XM_006712532.1
XP_006712595.1
17
6087
XM_011511186.1
XP_011509488.1
17
5997
XM_011511187.1
XP_011509489.1
16
5891
XM_011511188.1
XP_011509490.1
16
5891
XM_011511190.1
XP_011509492.1
15
5801
XM_011511191.1
XP_011509493.1
15
5759
XM_011511192.1
XP_011509494.1
15
5723
XM_011511193.1
XP_011509495.1
15
5720
XM_011511195.1
XP_011509497.1
15
5645
XM_011511196.2
XP_011509498.1
16
5832
XM_011511197.2
XP_011509499.1
16
5829
XM_017004111.1
XP_016859600.1
15
5720
XM_017004112.1
XP_016859601.1
16
5844
XM_017004113.1
XP_016859602.1
16
5736
XM_017004114.1
XP_016859603.1
15
5673
XM_017004115.1
XP_016859604.1
14
5465
XM_017004116.1
XP_016859605.1
14
5462
XM_017004117.1
XP_016859606.1
14
5462
XM_017004118.1
XP_016859607.1
14
5487
XM_017004119.1
XP_016859608.1
14
5577
XM_017004120.1
XP_016859609.1
14
5577
XM_017004121.1
XP_016859610.1
14
5484
XM_017004122.1
XP_016859611.1
14
5484
XM_017004123.1
XP_016859612.1
14
5568
XM_017004124.1
XP_016859613.1
14
5475
XM_017004125.1
XP_016859614.1
14
5565
XM_017004126.1
XP_016859615.1
14
5565
XM_017004127.1
XP_016859616.1
14
5565
XM_017004128.1
XP_016859617.1
13
5366
XM_017004129.1
XP_016859618.1
13
5363
XM_017004130.1
XP_016859619.1
13
5816
XM_017004131.1
XP_016859620.1
12
5208
XM_017004132.1
XP_016859621.1
12
5196
XM_017004133.1
XP_016859622.1
11
5115
XM_017004134.1
XP_016859623.1
11
5103
XM_017004135.1
XP_016859624.1
11
5549
XM_017004136.1
XP_016859625.1
11
5112
XM_017004137.1
XP_016859626.1
11
5100
XM_017004138.1
XP_016859627.1
13
1683
XM_017004139.1
XP_016859628.1
10
1440
XM_017004140.1
XP_016859629.1
11
1419
GenBank mRNA
AB209330.1
-
10
1311
AK291446.1
-
8
1182
AL833567.1
-
5
878
BC027583.1
-
11
1465
BC038857.1
-
14
1849
BC110423.1
-
11
1641
BC117123.1
-
9
1222
BC143245.1
-
9
1227
S76756.1
-
5
627
U89329.1
-
8
1148
U89330.1
-
11
5204
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-wl7lb:80/10.239.167.53:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0