Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144388
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
15
2743
NM_002374.3
NP_002365.3
15
6344
NM_031845.2
NP_114033.2
12
2276
NM_031847.2
NP_114035.2
13
2369
XM_005246555.1
XP_005246612.1
18
6973
XM_005246565.1
XP_005246622.1
14
6262
XM_005246566.1
XP_005246623.1
14
6250
XM_006712532.1
XP_006712595.1
18
6970
XM_011511186.1
XP_011509488.1
18
6880
XM_011511187.1
XP_011509489.1
17
6774
XM_011511188.1
XP_011509490.1
17
6774
XM_011511190.1
XP_011509492.1
16
6684
XM_011511191.1
XP_011509493.1
16
6642
XM_011511192.1
XP_011509494.1
16
6606
XM_011511193.1
XP_011509495.1
16
6603
XM_011511195.1
XP_011509497.1
16
6528
XM_011511196.2
XP_011509498.1
17
6715
XM_011511197.2
XP_011509499.1
17
6712
XM_017004111.1
XP_016859600.1
16
6603
XM_017004112.1
XP_016859601.1
17
6727
XM_017004113.1
XP_016859602.1
17
6619
XM_017004114.1
XP_016859603.1
16
6556
XM_017004115.1
XP_016859604.1
15
6348
XM_017004116.1
XP_016859605.1
15
6345
XM_017004117.1
XP_016859606.1
15
6345
XM_017004118.1
XP_016859607.1
15
6370
XM_017004119.1
XP_016859608.1
15
6460
XM_017004120.1
XP_016859609.1
15
6460
XM_017004121.1
XP_016859610.1
15
6367
XM_017004122.1
XP_016859611.1
15
6367
XM_017004123.1
XP_016859612.1
15
6451
XM_017004124.1
XP_016859613.1
15
6358
XM_017004125.1
XP_016859614.1
15
6448
XM_017004126.1
XP_016859615.1
15
6448
XM_017004127.1
XP_016859616.1
15
6448
XM_017004128.1
XP_016859617.1
14
6249
XM_017004129.1
XP_016859618.1
14
6246
XM_017004130.1
XP_016859619.1
14
6699
XM_017004131.1
XP_016859620.1
13
6091
XM_017004132.1
XP_016859621.1
13
6079
XM_017004133.1
XP_016859622.1
12
5998
XM_017004134.1
XP_016859623.1
12
5986
XM_017004135.1
XP_016859624.1
12
6432
XM_017004136.1
XP_016859625.1
12
5995
XM_017004137.1
XP_016859626.1
12
5983
XM_017004138.1
XP_016859627.1
14
2566
XM_017004139.1
XP_016859628.1
11
2323
XM_017004140.1
XP_016859629.1
12
2302
GenBank mRNA
AB209330.1
-
11
2194
AF088065.1
-
1
227
AL833567.1
-
6
1761
AV718243.1
-
1
531
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0