Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD46 SIRNA Assays ›
Gene Symbol:
CD46
Gene Name:
CD46 molecule
Gene Aliases:
AHUS2, MCP, MIC10, TLX, TRA2.10
Chromosome Location:
Chr.1: 207752038 - 207795516 on Build GRCh38
UniGene ID:
Hs.510402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144430
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD46
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002389.4
NP_002380.3
2
298
NM_153826.3
NP_722548.1
2
298
NM_172350.2
NP_758860.1
2
298
NM_172351.2
NP_758861.1
2
298
NM_172352.2
NP_758862.1
2
298
NM_172353.2
NP_758863.1
2
298
NM_172359.2
NP_758869.1
2
298
NM_172361.2
NP_758871.1
2
298
XM_011509563.1
XP_011507865.1
2
298
XM_011509564.1
XP_011507866.1
2
298
XM_017001308.1
XP_016856797.1
2
298
XM_017001309.1
XP_016856798.1
2
298
XM_017001310.1
XP_016856799.1
2
298
GenBank mRNA
A18585.1
-
2
156
AF209712.1
-
2
123
AF209713.1
-
2
123
AF209714.1
-
2
123
AK222822.1
-
2
224
AK291227.1
-
2
252
AK296819.1
-
2
250
AU131238.2
-
2
279
BC030594.2
-
2
238
4823040
BX537451.1
-
2
269
BX640613.1
-
2
280
BX649050.1
-
2
269
D84105.1
-
2
144
DB078198.1
-
2
298
EF076055.1
-
2
156
EF076056.1
-
2
156
EF076057.1
-
2
156
EF076058.1
-
2
156
M58050.1
-
2
157
S51940.1
-
2
210
X59405.1
-
2
268
X59406.1
-
2
227
X59407.1
-
2
227
X59408.1
-
2
268
X59409.1
-
2
227
X59410.1
-
2
227
Y00651.1
-
2
166
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell mediated immunity
proteolysis
complement activation, classical pathway
single fertilization
regulation of Notch signaling pathway
positive regulation of gene expression
negative regulation of gene expression
regulation of complement activation
interleukin-10 production
positive regulation of interleukin-10 production
sequestering of extracellular ligand from receptor
positive regulation of T cell proliferation
negative regulation of catalytic activity
positive regulation of memory T cell differentiation
innate immune response
positive regulation of regulatory T cell differentiation
negative regulation of complement activation
viral entry into host cell
positive regulation of transforming growth factor beta production
virus receptor activity
complement binding
endopeptidase activity
enzyme inhibitor activity
receptor activity
protein binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0