Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144434
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
3
295
NM_001018017.2
NP_001018017.1
3
268
NM_001044390.2
NP_001037855.1
3
249
NM_001044391.2
NP_001037856.1
3
268
NM_001044392.2
NP_001037857.1
3
295
NM_001044393.2
NP_001037858.1
3
249
NM_001204285.1
NP_001191214.1
3
928
NM_001204286.1
NP_001191215.1
3
955
NM_001204287.1
NP_001191216.1
3
349
NM_001204288.1
NP_001191217.1
3
295
NM_001204292.1
NP_001191221.1
3
276
NM_001204293.1
NP_001191222.1
3
322
NM_001204294.1
NP_001191223.1
3
249
NM_001204295.1
NP_001191224.1
3
295
NM_001204296.1
NP_001191225.1
3
276
NM_001204297.1
NP_001191226.1
3
349
NM_002456.5
NP_002447.4
3
322
GenBank mRNA
AF125525.1
-
3
187
AF348143.1
-
3
274
AK301575.1
-
4
1180
AY327582.1
-
3
201
AY327583.1
-
3
220
AY327584.1
-
3
247
AY327587.1
-
3
273
AY327590.1
-
3
201
AY327591.1
-
3
201
AY327592.1
-
3
220
AY327595.1
-
3
220
AY327596.1
-
4
403
AY327597.1
-
3
201
AY327598.1
-
3
201
AY327599.1
-
3
228
AY327600.1
-
3
220
AY466157.1
-
3
177
BC120974.2
-
3
245
40115473
BC120975.2
-
3
272
40115475
EF583642.1
-
3
598
EF583643.1
-
3
426
EF583644.1
-
3
412
EF583645.1
-
3
372
EF583647.1
-
3
327
EF583648.1
-
3
254
EF583649.1
-
3
273
EF583650.1
-
3
210
EF583651.1
-
3
210
EF583655.1
-
3
273
EF670706.1
-
3
622
EF670707.1
-
3
291
EF670709.1
-
3
254
EF670710.1
-
3
246
EF670711.1
-
3
300
EF670712.1
-
3
227
EF670713.1
-
3
273
EF670716.1
-
3
273
EF670717.1
-
3
273
EF670718.1
-
3
327
EF670719.1
-
3
297
EF670721.1
-
3
254
FJ226028.1
-
2
729
FJ226029.1
-
2
887
FJ226030.1
-
3
857
FJ226031.1
-
2
815
FJ226032.1
-
2
748
FJ226033.1
-
2
729
FJ226034.1
-
3
648
FJ226036.1
-
2
748
FJ226037.1
-
3
426
FJ226038.1
-
2
747
FJ226040.1
-
3
273
FJ226042.1
-
3
327
FJ226043.1
-
3
313
FJ226044.1
-
3
353
FJ226046.1
-
3
399
FJ226047.1
-
3
227
FJ226049.1
-
3
327
FJ226050.1
-
3
300
FJ226052.1
-
3
246
FJ226053.1
-
3
254
FJ226054.1
-
3
372
FJ226055.1
-
3
254
J05581.1
-
3
928
J05582.1
-
3269
M34088.1
-
1
415
M34089.1
-
1
415
U60259.1
-
3
202
U60260.1
-
3
221
U60261.1
-
3
275
X52228.1
-
4
1033
X52229.1
-
4
1033
X80761.1
-
3
196
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline