Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144435
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
5
396
NM_001018017.2
NP_001018017.1
5
369
NM_001204285.1
NP_001191214.1
5
1029
NM_001204286.1
NP_001191215.1
5
1056
NM_001204287.1
NP_001191216.1
5
450
NM_001204288.1
NP_001191217.1
5
396
NM_001204289.1
NP_001191218.1
4
318
NM_001204290.1
NP_001191219.1
4
255
NM_001204291.1
NP_001191220.1
4
327
NM_001204292.1
NP_001191221.1
4
321
NM_001204293.1
NP_001191222.1
5
423
NM_001204294.1
NP_001191223.1
4
294
NM_002456.5
NP_002447.4
5
423
GenBank mRNA
AF125525.1
-
5
288
AK301575.1
-
6
1281
AY327587.1
-
5
374
AY327588.1
-
4
269
AY327596.1
-
6
504
AY466157.1
-
5
278
BC120974.2
-
5
346
40115473
BC120975.2
-
5
373
40115475
EF583642.1
-
5
699
EF583643.1
-
5
527
EF583644.1
-
5
513
EF583645.1
-
5
473
EF583646.1
-
5
441
EF583647.1
-
5
428
EF583648.1
-
5
355
EF583649.1
-
5
374
EF583651.1
-
4, 5
306
EF583652.1
-
4
296
EF583653.1
-
4
254
EF583654.1
-
4
233
EF670706.1
-
5
723
EF670707.1
-
5
392
EF670708.1
-
4
305
EF670709.1
-
4
299
EF670710.1
-
4
291
EF670711.1
-
5
401
EF670712.1
-
4
272
EF670714.1
-
4
227
FJ226028.1
-
4
830
FJ226029.1
-
4
988
FJ226030.1
-
5
958
FJ226031.1
-
4
916
FJ226032.1
-
4
849
FJ226033.1
-
4
830
FJ226034.1
-
5
749
FJ226037.1
-
5
527
FJ226039.1
-
5
454
FJ226040.1
-
5
374
FJ226041.1
-
6
531
FJ226042.1
-
5
428
FJ226043.1
-
5
414
FJ226044.1
-
4
398
FJ226045.1
-
4
368
FJ226047.1
-
5
328
FJ226052.1
-
5
347
J05581.1
-
5
1029
J05582.1
-
3370
M34088.1
-
3
516
M34089.1
-
3
516
U60259.1
-
4
247
U60260.1
-
5
322
U60261.1
-
5
376
X52228.1
-
4
1383
X52229.1
-
6
1134
X80761.1
-
5
297
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0