Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144436
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
5
489
NM_001018017.2
NP_001018017.1
5
462
NM_001204285.1
NP_001191214.1
5
1122
NM_001204286.1
NP_001191215.1
5
1149
NM_001204287.1
NP_001191216.1
5
543
NM_001204288.1
NP_001191217.1
5
489
NM_001204289.1
NP_001191218.1
4
411
NM_001204290.1
NP_001191219.1
4
348
NM_001204291.1
NP_001191220.1
4
420
NM_001204292.1
NP_001191221.1
4
414
NM_001204293.1
NP_001191222.1
5
516
NM_001204294.1
NP_001191223.1
4
387
NM_002456.5
NP_002447.4
5
516
GenBank mRNA
AF125525.1
-
5
381
AK301575.1
-
6
1374
AY327587.1
-
5
467
AY327588.1
-
4
362
AY327596.1
-
6
597
BC120974.2
-
5
439
40115473
BC120975.2
-
5
466
40115475
EF583642.1
-
5
792
EF583643.1
-
5
620
EF583644.1
-
5
606
EF583645.1
-
5
566
EF583646.1
-
5
534
EF583647.1
-
5
521
EF583648.1
-
5
448
EF583649.1
-
5
467
EF583650.1
-
5
404
EF583651.1
-
5
399
EF583653.1
-
4
347
EF583654.1
-
4
326
EF670706.1
-
5
816
EF670707.1
-
5
485
EF670708.1
-
4
398
EF670709.1
-
4
392
EF670710.1
-
4
384
EF670711.1
-
5
494
EF670712.1
-
4
365
EF670714.1
-
4
320
FJ226028.1
-
4
923
FJ226029.1
-
4
1081
FJ226030.1
-
5
1051
FJ226031.1
-
4
1009
FJ226032.1
-
4
942
FJ226033.1
-
4
923
FJ226034.1
-
5
842
FJ226037.1
-
5
620
FJ226039.1
-
5
547
FJ226040.1
-
5
467
FJ226041.1
-
6
624
FJ226042.1
-
5
521
FJ226043.1
-
5
507
FJ226044.1
-
4
491
FJ226045.1
-
4
461
FJ226047.1
-
5
421
FJ226052.1
-
5
440
J05581.1
-
5
1122
J05582.1
-
3463
M34088.1
-
3
609
M34089.1
-
3
609
U60259.1
-
4
340
U60260.1
-
5
415
U60261.1
-
5
469
X52228.1
-
4
1476
X52229.1
-
6
1227
X80761.1
-
5
390
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline