Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136024.3 | NP_001129496.1 | 2 | 345 | ||
NM_001291867.1 | NP_001278796.1 | 2 | 940 | |||
NM_001291868.1 | NP_001278797.1 | 2 | 345 | |||
NM_198270.3 | NP_938011.1 | 2 | 940 | |||
GenBank mRNA | AK302437.1 | - | 2 | 1126 | ||
AY436752.1 | - | 2 | 940 | |||
AY456992.1 | - | 2 | 345 | |||
AY456993.1 | - | 1 | 1000 | |||
BC136415.1 | - | 2 | 666 | 9020403 | ||
BC171763.1 | - | 2 | 666 | 9054416 | ||
CR936788.1 | - | 2 | 114 | |||
GQ988776.1 | - | 2 | 602 |