Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000613.2 | NP_000604.1 | 9 | 1065 | ||
GenBank mRNA | AK313648.1 | - | 9 | 1028 | ||
BC005395.1 | - | 9 | 1046 | 4071616 | ||
J03048.1 | - | 9 | 1001 | |||
X02537.1 | - | 9 | 941 |