Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000621.4 | NP_000612.1 | 4 | 1859 | ||
NM_001165947.2 | NP_001159419.1 | 3 | 1119 | |||
GenBank mRNA | A23337.1 | - | 3 | 1335 | ||
AF498982.1 | - | 3 | 1128 | |||
AK302787.1 | - | 3 | 958 | |||
AK314132.1 | - | 4 | 1616 | |||
BC069356.1 | - | 3 | 1207 | 7262350 | ||
BC069576.1 | - | 3 | 1207 | 7262362 | ||
BC074848.2 | - | 3 | 1155 | 30915554 | ||
BC074849.2 | - | 3 | 1155 | 30915332 | ||
BC096839.1 | - | 3 | 1129 | 6519217 | ||
BX647777.1 | - | 3 | 903 | |||
M86841.1 | - | 3 | 1104 | |||
S71229.1 | - | 3 | 1178 | |||
X57830.1 | - | 3 | 1273 |
Set Membership: |
G-protein coupled receptor |