Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000818.2 | NP_000809.1 | 17 | 2427 | ||
GenBank mRNA | BP392604.1 | - | 1 | 95 | ||
M74826.1 | - | 17 | 2065 |