Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000861.3 | NP_000852.1 | 3 | 1075 | ||
NM_001098211.1 | NP_001091681.1 | 2 | 996 | |||
NM_001098212.1 | NP_001091682.1 | 2 | 946 | |||
NM_001098213.1 | NP_001091683.1 | 2 | 1226 | |||
XM_011533652.1 | XP_011531954.1 | 4 | 1363 | |||
XM_011533653.2 | XP_011531955.1 | 2 | 1192 | |||
XM_017006283.1 | XP_016861772.1 | 2 | 1146 | |||
XM_017006284.1 | XP_016861773.1 | 2 | 1185 | |||
GenBank mRNA | AF026261.1 | - | 1 | 884 | ||
AK289412.1 | - | 2 | 1226 | |||
AK292157.1 | - | 2 | 943 | |||
AK312879.1 | - | 2 | 944 | |||
AY136743.1 | - | 1 | 884 | |||
BC060802.1 | - | 3 | 1075 | 30340368 | ||
D28481.1 | - | 1 | 887 | |||
Z34897.1 | - | 1 | 884 |
Set Membership: |
![]() |