Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GDA SIRNA Assays ›
Gene Symbol:
GDA
Gene Name:
guanine deaminase
Gene Aliases:
CYPIN, GUANASE, NEDASIN
Chromosome Location:
Chr.9: 72114595 - 72256045 on Build GRCh38
UniGene ID:
Hs.494163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14471
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GDA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242505.2
NP_001229434.1
3, 4
603
NM_001242506.2
NP_001229435.1
4, 5
644
NM_001242507.2
NP_001229436.1
3, 4
510
NM_004293.4
NP_004284.1
3, 4
603
XM_005252317.1
XP_005252374.1
3, 4
601
XM_011519213.1
XP_011517515.1
4, 5
715
XM_011519214.1
XP_011517516.1
4, 5
715
XM_011519215.1
XP_011517517.1
4, 5
715
XM_011519216.1
XP_011517518.1
4, 5
715
XM_011519217.1
XP_011517519.1
4, 5
715
XM_011519218.2
XP_011517520.1
4, 5
600
XM_011519219.1
XP_011517521.1
4, 5
713
XM_011519220.1
XP_011517522.1
3, 4
599
XM_011519221.1
XP_011517523.1
3, 4
510
XM_011519222.1
XP_011517524.1
4, 5
715
XM_017015333.1
XP_016870822.1
4, 5
715
XM_017015334.1
XP_016870823.1
4, 5
715
XM_017015335.1
XP_016870824.1
4, 5
715
XM_017015336.1
XP_016870825.1
3, 4
601
XM_017015337.1
XP_016870826.1
3, 4
601
XM_017015338.1
XP_016870827.1
5, 6
671
XM_017015339.1
XP_016870828.1
3, 4
599
GenBank mRNA
AB033084.1
-
3, 4
470
AF019638.1
-
3, 4
479
AF095286.1
-
3, 4
487
AK097540.1
-
4, 5
702
AK300418.1
-
3, 4
510
AK315988.1
-
4, 5
422
BC053584.1
-
3, 4
577
5186337
Back To Top
More Information
Panther Classification:
Molecular Function -
deaminase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
guanine catabolic process
purine nucleotide catabolic process
nervous system development
guanine metabolic process
zinc ion binding
guanine deaminase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0