Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002023.4 | NP_002014.2 | 3 | 2501 | ||
NR_103757.1 | 2 | 1515 | ||||
GenBank mRNA | AK092899.1 | - | 4 | 1962 | ||
AK098322.1 | - | 2 | 1575 | |||
BC035281.1 | - | 3 | 2177 | 5181410 | ||
U05291.1 | - | 2 | 1144 |