Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002077.3 | NP_002068.1 | 23 | 4315 | ||
XM_005251929.3 | XP_005251986.1 | 23 | 4247 | |||
XM_006717062.3 | XP_006717125.1 | 23 | 4311 | |||
XM_006717063.3 | XP_006717126.1 | 23 | 4282 | |||
GenBank mRNA | AB208858.1 | - | 18 | 3666 | ||
BC032853.1 | - | 23 | 4285 | 4828793 | ||
U51587.1 | - | 23 | 4305 |